accessibility menu, dialog, popup

UserName

CAS RN 76387-70-7

H3CCH3CH3OONHNH2OHO

CAS RN 76387-70-7

IUPAC Name: 3-amino-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 3-amino-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 204.23
Molecular Formula: C8H16N2O4
InChi Key: KRJLRVZLNABMAT-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CN)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results