CAS RN 7625-01-6
CAS RN 7625-01-6
IUPAC Name:
ethyl 3-methyl-4H-1,4-benzothiazine-2-carboxylate
Synonyms:
ethyl 3-methyl-4H-1,4-benzothiazine-2-carboxylate
Molecular Weight (g/mol):
235.3
Molecular Formula:
C12H13NO2S
InChi Key:
NCZIRXVLKKERGP-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(C)NC2=CC=CC=C2S1
1
–
1
of
1
results
1
Ethyl 3-methyl-4H-1,4-benzothiazine-2-carboxylate, 98%, Thermo Scientific™
CAS: 7625-01-6 MDL Number: MFCD00173950 PubChem CID: 360308 IUPAC Name: ethyl 3-methyl-4H-1,4-benzothiazine-2-carboxylate SMILES: CCOC(=O)C1=C(NC2=CC=CC=C2S1)C