accessibility menu, dialog, popup

UserName

HNNF

CAS RN 76141-89-4

IUPAC Name: 1-[(4-fluorophenyl)methyl]-1,4-diazepane
Synonyms: 1-[(4-fluorophenyl)methyl]-1,4-diazepane
Molecular Weight (g/mol): 208.28
Molecular Formula: C12H17FN2
InChi Key: KZCHYFUMQQWGJK-UHFFFAOYSA-N
SMILES: FC1=CC=C(CN2CCCNCC2)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)