accessibility menu, dialog, popup

UserName

H3CNNON

CAS RN 76081-98-6

IUPAC Name: 2-[8-(benzyloxy)-2-methylimidazo[1,2-a]pyridin-3-yl]acetonitrile
Synonyms: 2-[8-(benzyloxy)-2-methylimidazo[1,2-a]pyridin-3-yl]acetonitrile
Molecular Weight (g/mol): 277.33
Molecular Formula: C17H15N3O
InChi Key: PYKJFEPAUKAXNN-UHFFFAOYSA-N
SMILES: CC1=C(CC#N)N2C=CC=C(OCC3=CC=CC=C3)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results