accessibility menu, dialog, popup

UserName

OHClOOO

CAS RN 7601-90-3

IUPAC Name: perchloric acid
Synonyms: perchloric acid perchlorsaeure acids, perchloric perchloric acid (substance) ueberchlorsaeure ClHO4 perchloric acids HClO4 acid, perchloric perchlorates
Molecular Weight (g/mol): 100.45
Molecular Formula: ClHO4
InChi Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES: O[Cl](=O)(=O)=O
Quantity 
  • (25)
  • (1)
  • (7)
  • (1)
  • (12)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (17)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (66)
  • (6)
Grade 
  • (3)
  • (14)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (15)
  • (24)
  • (1)
  • (1)
  • (1)
  • (8)
  • (9)
  • (16)
  • (1)
  • (25)
  • (1)
  • (7)
  • (1)
  • (12)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (17)
  • (1)
  • (1)
  • (66)
  • (6)
  • (3)
  • (14)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)