accessibility menu, dialog, popup

UserName

CAS RN 760-93-0

H3CCH2OOOCH3CH2

CAS RN 760-93-0

IUPAC Name: 2-methylprop-2-enoyl 2-methylprop-2-enoate
Synonyms: methacrylic anhydride
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: DCUFMVPCXCSVNP-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC(=O)C(C)=C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)