accessibility menu, dialog, popup

UserName

CAS RN 76-43-7

IUPAC Name: (1S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-1,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: fluoxymesterone android F android-F android-f ora-testryl ora testryl androsterolo fluoximesterona [inn-spanish] FXM 氟甲睾酮
Molecular Weight (g/mol): 336.45
Molecular Formula: C20H29FO3
InChi Key: YLRFCQOZQXIBAB-RBZZARIASA-N
SMILES: C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Certifications/Compliance 
  • (2)
  • (1)
Industry Type 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results