CAS RN 76-05-1
Trifluoroacetic Acid, 99+%, for HPLC, Fisher Chemical™
CAS: 76-05-1 Molecular Formula: C2HF3O2 Molecular Weight (g/mol): 114.02 MDL Number: MFCD00004169 InChI Key: DTQVDTLACAAQTR-UHFFFAOYSA-N Synonym: trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoroacetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova PubChem CID: 6422 ChEBI: CHEBI:45892 SMILES: OC(=O)C(F)(F)F
Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
CAS: 76-05-1 Molecular Formula: C2HF3O2 Molecular Weight (g/mol): 114.02 MDL Number: MFCD00004169 InChI Key: DTQVDTLACAAQTR-UHFFFAOYSA-N Synonym: trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoroacetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova PubChem CID: 6422 ChEBI: CHEBI:45892 IUPAC Name: trifluoroacetic acid SMILES: OC(=O)C(F)(F)F
Trifluoroacetic acid, 99+%, extra pure
CAS: 76-05-1 Molecular Formula: C2HF3O2 Molecular Weight (g/mol): 114.02 MDL Number: MFCD00004169 InChI Key: DTQVDTLACAAQTR-UHFFFAOYSA-N Synonym: trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoroacetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova PubChem CID: 6422 ChEBI: CHEBI:45892 SMILES: OC(=O)C(F)(F)F
Trifluoroacetic acid, 99+%, for HPLC
CAS: 76-05-1 Molecular Formula: C2HF3O2 Molecular Weight (g/mol): 114.02 MDL Number: MFCD00004169 InChI Key: DTQVDTLACAAQTR-UHFFFAOYSA-N Synonym: trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoroacetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova PubChem CID: 6422 ChEBI: CHEBI:45892 IUPAC Name: 2,2,2-trifluoroacetic acid SMILES: OC(=O)C(F)(F)F
Trifluoroacetic acid, 99.5%, for biochemistry
CAS: 76-05-1 Molecular Formula: C2HF3O2 Molecular Weight (g/mol): 114.02 MDL Number: MFCD00004169 InChI Key: DTQVDTLACAAQTR-UHFFFAOYSA-N Synonym: trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoroacetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova PubChem CID: 6422 ChEBI: CHEBI:45892 IUPAC Name: trifluoroacetic acid SMILES: OC(=O)C(F)(F)F
Trifluoroacetic acid, HPLC Grade, 99.5+%
CAS: 76-05-1 Molecular Formula: C2HF3O2 Molecular Weight (g/mol): 114.02 MDL Number: MFCD00004169 InChI Key: DTQVDTLACAAQTR-UHFFFAOYSA-N Synonym: trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoroacetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova PubChem CID: 6422 ChEBI: CHEBI:45892 IUPAC Name: 2,2,2-trifluoroacetic acid SMILES: OC(=O)C(F)(F)F
Trifluoroacetic acid, 98%, Thermo Scientific™
CAS: 76-05-1 Molecular Formula: C2HF3O2 Molecular Weight (g/mol): 114.02 MDL Number: MFCD00004169 InChI Key: DTQVDTLACAAQTR-UHFFFAOYSA-N Synonym: TFA IUPAC Name: trifluoroacetic acid SMILES: OC(=O)C(F)(F)F
Acetonitrile with 0.05% Trifluoroacetic Acid (TFA), Optima LC/MS, Fisher Chemical
Acetonitrile with 0.05% TFA, LC/MS Grade, Thermo Scientific
Trifluoroacetic acid, 99%
CAS: 76-05-1 Molecular Formula: C2HF3O2 Molecular Weight (g/mol): 114.02 MDL Number: MFCD00004169 InChI Key: DTQVDTLACAAQTR-UHFFFAOYSA-N Synonym: trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoroacetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova PubChem CID: 6422 ChEBI: CHEBI:45892 IUPAC Name: trifluoroacetic acid SMILES: OC(=O)C(F)(F)F
Trifluoroacetic acid, 99.5+%
CAS: 76-05-1 Molecular Formula: C2HF3O2 Molecular Weight (g/mol): 114.02 MDL Number: MFCD00004169 InChI Key: DTQVDTLACAAQTR-UHFFFAOYSA-N Synonym: trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoroacetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova PubChem CID: 6422 ChEBI: CHEBI:45892 IUPAC Name: trifluoroacetic acid SMILES: OC(=O)C(F)(F)F
Thermo Scientific Chemicals Trifluoroacetic acid, biochemical grade, 99.5+%
CAS: 76-05-1 Molecular Formula: C2HF3O2 Molecular Weight (g/mol): 114.02 MDL Number: MFCD00004169 InChI Key: DTQVDTLACAAQTR-UHFFFAOYSA-N Synonym: trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoroacetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova PubChem CID: 6422 ChEBI: CHEBI:45892 IUPAC Name: trifluoroacetic acid SMILES: OC(=O)C(F)(F)F