accessibility menu, dialog, popup

UserName

H3COOCH3H3COOCH3

CAS RN 759-36-4

IUPAC Name: 1,3-diethyl 2-(propan-2-yl)propanedioate
Synonyms: 1,3-diethyl 2-isopropylpropanedioate
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: BYQFBFWERHXONI-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C(C)C)C(=O)OCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)