CAS RN 758-17-8
CAS RN 758-17-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Methyl-N-formylhydrazine, TRC
CAS: 758-17-8 Molecular Formula: C2 H6 N2 O Molecular Weight (g/mol): 74.082 Synonym: 1-Formyl-1-methylhydrazine,1-Methyl-1-formylhydrazide,2-Methyl-2-formylhydrazine,N-Formyl-N-methylhydrazine,Formic Acid 1-Methylhydrazide,1-Methylhydrazinecarboxaldehyde IUPAC Name: N-amino-N-methyl-formamide SMILES: CN(N)C=O