accessibility menu, dialog, popup

UserName

NHOCl

CAS RN 7578-45-2

IUPAC Name: 4-chloro-N-phenylbutanamide
Synonyms: 4-chloro-N-phenylbutanamide
Molecular Weight (g/mol): 197.66
Molecular Formula: C10H12ClNO
InChi Key: GJMGKNWSRKDALN-UHFFFAOYSA-N
SMILES: ClCCCC(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results