CAS RN 7570-45-8
CAS RN 7570-45-8
IUPAC Name:
9-ethyl-9H-carbazole-3-carbaldehyde
Synonyms:
9-ethylcarbazole-3-carbaldehyde
Molecular Weight (g/mol):
223.28
Molecular Formula:
C15H13NO
InChi Key:
QGJXVBICNCIWEL-UHFFFAOYSA-N
SMILES:
CCN1C2=C(C=CC=C2)C2=C1C=CC(C=O)=C2
No Refinements
1
–
0
of
0
results
1
N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
CAS: 7570-45-8 Molecular Formula: C15H13NO Molecular Weight (g/mol): 223.28 MDL Number: MFCD00004963 InChI Key: QGJXVBICNCIWEL-UHFFFAOYSA-N Synonym: 9-ethyl-9h-carbazole-3-carbaldehyde,n-ethyl-3-carbazolecarboxaldehyde,n-ethylcarbazole-3-carboxaldehyde,3-formyl-n-ethylcarbazole,n-ethylcarbazole-3-carbaldehyde,9-ethyl-3-carbazolecarboxaldehyde,ccris 9052,n-ethyl carbazole-3-aldehyde,n-ethyl-carbazole-3-aldehyde,pubchem9556 PubChem CID: 82055 IUPAC Name: 9-ethylcarbazole-3-carbaldehyde SMILES: CCN1C2=C(C=CC=C2)C2=C1C=CC(C=O)=C2