accessibility menu, dialog, popup

UserName

CAS RN 7568-92-5

IUPAC Name: 2-amino-2-phenylethan-1-ol
Synonyms: 2-amino-2-phenylethanol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: IJXJGQCXFSSHNL-UHFFFAOYNA-N
SMILES: NC(CO)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)