accessibility menu, dialog, popup

UserName

CH3NHNH2NHClHCl

CAS RN 75614-89-0

IUPAC Name: 1-(1H-imidazol-5-yl)propan-2-amine dihydrochloride
Synonyms: α-methylhistamine dihydrochloride
Molecular Weight (g/mol): 198.09
Molecular Formula: C6H13Cl2N3
InChi Key: IZHCNQFUWDFPCW-UHFFFAOYNA-N
SMILES: Cl.Cl.CC(N)CC1=CN=CN1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)