accessibility menu, dialog, popup

UserName

CAS RN 75611-06-2

IUPAC Name: 2-(1-benzofuran-3-yl)ethan-1-ol
Synonyms: 2-(1-benzofuran-3-yl)ethanol
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: QBEGKDXITSPUTE-UHFFFAOYSA-N
SMILES: OCCC1=COC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results