accessibility menu, dialog, popup

UserName

CAS RN 7560-83-0

H3CN

CAS RN 7560-83-0

IUPAC Name: N-cyclohexyl-N-methylcyclohexanamine
Synonyms: N-cyclohexyl-N-methylcyclohexanamine
Molecular Weight (g/mol): 195.35
Molecular Formula: C13H25N
InChi Key: GSCCALZHGUWNJW-UHFFFAOYSA-N
SMILES: CN(C1CCCCC1)C1CCCCC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)