accessibility menu, dialog, popup

UserName

CH3NHN

CAS RN 7554-65-6

IUPAC Name: 4-methyl-1H-pyrazole
Synonyms: fomepizole fomepizol [inn-spanish] fomepizole preparation paladin brand of fomepizole antizol fomepizole (substance) orphan brand of fomepizole fomepizole (product) fomepizol fomepizolum
Molecular Weight (g/mol): 82.11
Molecular Formula: C4H6N2
InChi Key: RIKMMFOAQPJVMX-UHFFFAOYSA-N
SMILES: CC1=CNN=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (3)
Quantity 
  • (4)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)