accessibility menu, dialog, popup

UserName

NNHN

CAS RN 75415-03-1

IUPAC Name: 2-(1H-pyrazol-5-yl)pyridine
Synonyms: 2-(2H-pyrazol-3-yl)pyridine
Molecular Weight (g/mol): 145.17
Molecular Formula: C8H7N3
InChi Key: HKEWOTUTAYJWQJ-UHFFFAOYSA-N
SMILES: N1N=CC=C1C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results