CAS RN 75345-27-6
CAS RN 75345-27-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Polyquaternium 1, TRC
CAS: 75345-27-6 Molecular Formula: (C6H12ClN)n.C16H36Cl2N2O6 Molecular Weight (g/mol): 556.99 Synonym: α-[4-[Tris(2-hydroxyethyl)ammonio]-2-buten-1-yl]-ω-[tris(2-hydroxyethyl)ammonio]poly[(dimethyliminio)-2-butene-1,4-diyl Chloride] Chloride,Onamer M,Onyxsperse 12S,Polidronium Chloride,Polyquad,Polyquat 1; IUPAC Name: dimethyl-bis[(E)-4-[tris(2-hydroxyethyl)azaniumyl]but-2-enyl]azanium;trichloride SMILES: C[N+](C/C=C/C[N+](CCO)(CCO)CCO)(C/C=C/C[N+](CCO)(CCO)CCO)C.[Cl-].[Cl-].[Cl-]