accessibility menu, dialog, popup

UserName

CAS RN 7533-40-6

IUPAC Name: (2S)-1-hydroxy-4-methylpentan-2-aminium
Synonyms: (2S)-1-hydroxy-4-methylpentan-2-aminium
Molecular Weight (g/mol): 118.2
Molecular Formula: C6H16NO
InChi Key: VPSSPAXIFBTOHY-LURJTMIESA-O
SMILES: CC(C)C[C@H]([NH3+])CO
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results