accessibility menu, dialog, popup

UserName

CAS RN 75291-85-9

NBrClONH2

CAS RN 75291-85-9

IUPAC Name: 5-bromo-2-chloropyridine-3-carboxamide
Synonyms: 5-bromo-2-chloropyridine-3-carboxamide
Molecular Weight (g/mol): 235.47
Molecular Formula: C6H4BrClN2O
InChi Key: NFUOTGWWIKJNQL-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(Br)=CN=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results