accessibility menu, dialog, popup

UserName

H3CNONOCH3H2OH2OH2OH2OH2OH2OH2OH2ONaNa(E/Z)(E/Z)

CAS RN 75006-64-3

IUPAC Name: disodium oxido[3-(oxidoimino)butan-2-ylidene]amine octahydrate
Synonyms: disodium oxido[3-(oxidoimino)butan-2-ylidene]amine octahydrate
Molecular Weight (g/mol): 304.2
Molecular Formula: C4H22N2Na2O10
InChi Key: IJJDRGUPRATWQX-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-]
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)