accessibility menu, dialog, popup

UserName

CH3H3CCH3OHO

CAS RN 75-98-9

IUPAC Name: 2,2-dimethylpropanoic acid
Synonyms: pivalic acid kyselina pivalova acido pivalico pivalinsaeure tert-pentanoic acid acide 2,2-dimethylpropanoique versatic 5 acide pivalique
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: IUGYQRQAERSCNH-UHFFFAOYSA-N
SMILES: CC(C)(C)C(O)=O
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (2)
  • (8)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (4)
  • (2)