accessibility menu, dialog, popup

UserName

CAS RN 75-86-5

IUPAC Name: 2-hydroxy-2-methylpropanenitrile
Synonyms: acetone cyanohydrin 2-methyl-lactonitrile acetonkyanhydrin acetone-cyanohydrin rcra waste number P069 acetoncianhidrinei acetoncyanhydrin acetoncianidrina cyanohydrin 2-propanone acetonecyanhydrine
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: MWFMGBPGAXYFAR-UHFFFAOYSA-N
SMILES: CC(C)(O)C#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results