accessibility menu, dialog, popup

UserName

CH3SiCH3ClCl

CAS RN 75-78-5

IUPAC Name: dichlorodimethylsilane
Synonyms: dimethyldichlorosilane
Molecular Weight (g/mol): 129.06
Molecular Formula: C2H6Cl2Si
InChi Key: LIKFHECYJZWXFJ-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)Cl
Molecular Weight (g/mol) 
  • (5)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (6)
Boiling Point 
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (6)
  • (3)
  • (3)
  • (5)
  • (9)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (6)
  • (8)