accessibility menu, dialog, popup

UserName

CAS RN 75-66-1

CH3CH3CH3HS

CAS RN 75-66-1

IUPAC Name: 2-methylpropane-2-thiol
Synonyms: 2-methyl-2-propanethiol
Molecular Weight (g/mol): 90.18
Molecular Formula: C4H10S
InChi Key: WMXCDAVJEZZYLT-UHFFFAOYSA-N
SMILES: CC(C)(C)S
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)