accessibility menu, dialog, popup

UserName

CAS RN 75-47-8

IUPAC Name: triiodomethane
Synonyms: iodoform dezinfekt V jodoform ccris 346 trijodmethane 碘仿
Molecular Weight (g/mol): 393.73
Molecular Formula: CHI3
InChi Key: OKJPEAGHQZHRQV-UHFFFAOYSA-N
SMILES: IC(I)I
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)