accessibility menu, dialog, popup

UserName

CH3CH3HS

CAS RN 75-33-2

IUPAC Name: propane-2-thiol
Synonyms: 2-propanethiol
Molecular Weight (g/mol): 76.16
Molecular Formula: C3H8S
InChi Key: KJRCEJOSASVSRA-UHFFFAOYSA-N
SMILES: CC(C)S
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (6)