accessibility menu, dialog, popup

UserName

CAS RN 75-22-9

NCH3H3CCH3BH3

CAS RN 75-22-9

IUPAC Name: boranyltrimethylazanium
Synonyms: boranyltrimethylazanium
Molecular Weight (g/mol): 71.94
Molecular Formula: C3H11BN
InChi Key: BNMCRXUHAWPWPT-UHFFFAOYSA-N
SMILES: B[N+](C)(C)C
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)