accessibility menu, dialog, popup

UserName

CH3SnH3CCH3H2C

CAS RN 7486-35-3

IUPAC Name: tributyl(ethenyl)stannane
Synonyms: tributyl(ethenyl)stannane
Molecular Weight (g/mol): 317.1
Molecular Formula: C14H30Sn
InChi Key: QIWRFOJWQSSRJZ-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C=C
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (6)
  • (3)
  • (3)
  • (3)