CAS RN 74848-84-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Thymidine-d3, TRC
CAS: 74848-84-3 Molecular Formula: C10 2H3 H11 N2 O5 Molecular Weight (g/mol): 245.25 Synonym: Thymidine D3 IUPAC Name: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trideuteriomethyl)pyrimidine-2,4-dione SMILES: [2H]C([2H])([2H])C1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O
2
Thymidine-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.