accessibility menu, dialog, popup

UserName

CAS RN 7461-50-9

NH2NClN

CAS RN 7461-50-9

IUPAC Name: 2-chloropyrimidin-4-amine
Synonyms: 2-chloropyrimidin-4-amine
Molecular Weight (g/mol): 129.55
Molecular Formula: C4H4ClN3
InChi Key: LPBDZVNGCNTELM-UHFFFAOYSA-N
SMILES: NC1=CC=NC(Cl)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results