accessibility menu, dialog, popup

UserName

H3COOCH3CH3

CAS RN 7452-79-1

IUPAC Name: ethyl 2-methylbutanoate
Synonyms: ethyl 2-methylbutyrate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: HCRBXQFHJMCTLF-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)CC
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results