accessibility menu, dialog, popup

UserName

CAS RN 7450-57-9

NH2HNOOHH2N

CAS RN 7450-57-9

IUPAC Name: 3-amino-4-hydroxybenzohydrazide
Synonyms: 3-amino-4-hydroxybenzohydrazide
Molecular Weight (g/mol): 167.17
Molecular Formula: C7H9N3O2
InChi Key: KNKNIBYXWQUDIH-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(O)C(N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results