accessibility menu, dialog, popup

UserName

PbOSOOO

CAS RN 7446-14-2

IUPAC Name: λ²-lead(2+) sulfate
Synonyms: lead sulfate
Molecular Weight (g/mol): 303.3
Molecular Formula: O4PbS
InChi Key: KEQXNNJHMWSZHK-UHFFFAOYSA-L
SMILES: [Pb++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Physical Form 
  • (2)
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Grade 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (6)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)