CAS RN 7440-65-5
Yttrium pieces, 99.9% (REO)
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N Synonym: yttrium, elemental,yttrium-89,ion,ytrio,hydride,unii-58784xqc3y,yttrium, metal and compounds,ingot,foil,element PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium foil, 0.25mm (0.01in) thick, 99.9% (REO)
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N Synonym: yttrium, elemental,yttrium-89,ion,ytrio,hydride,unii-58784xqc3y,yttrium, metal and compounds,ingot,foil,element PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N Synonym: yttrium, elemental,yttrium-89,ion,ytrio,hydride,unii-58784xqc3y,yttrium, metal and compounds,ingot,foil,element PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium foil, 0.025mm (0.001in) thick, 99.9% (REO)
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N Synonym: yttrium, elemental,yttrium-89,ion,ytrio,hydride,unii-58784xqc3y,yttrium, metal and compounds,ingot,foil,element PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium foil, 0.127mm (0.005in) thick, 99.9% (REO)
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N Synonym: yttrium, elemental,yttrium-89,ion,ytrio,hydride,unii-58784xqc3y,yttrium, metal and compounds,ingot,foil,element PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium foil, 0.64mm (0.025in) thick, 99.5% (REO)
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N Synonym: yttrium, elemental,yttrium-89,ion,ytrio,hydride,unii-58784xqc3y,yttrium, metal and compounds,ingot,foil,element PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium ingot, 99.9% (REO)
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N Synonym: yttrium, elemental,yttrium-89,ion,ytrio,hydride,unii-58784xqc3y,yttrium, metal and compounds,ingot,foil,element PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium foil, 0.1mm (0.004in) thick, 99.6% (REO)
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N Synonym: yttrium, elemental,yttrium-89,ion,ytrio,hydride,unii-58784xqc3y,yttrium, metal and compounds,ingot,foil,element PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N Synonym: yttrium, elemental,yttrium-89,ion,ytrio,hydride,unii-58784xqc3y,yttrium, metal and compounds,ingot,foil,element PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium powder, -40 mesh, 99.6% (REO)
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N Synonym: yttrium, elemental,yttrium-89,ion,ytrio,hydride,unii-58784xqc3y,yttrium, metal and compounds,ingot,foil,element PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium Standard, 2% HNO3 in 1000mg/L (Assurance Grade), SPEX CertiPrep™
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium Standard For ICP, 2% HNO3 in 1000μg/mL, Assurance Grade, SPEX CertiPrep™
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.91 MDL Number: MFCD00011468 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Yttrium Standard: Y @ 5000 μg/g in Hydrocarbon Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Yttrium Standard: Y @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2