CAS RN 7440-55-3
Gallium, 99.99%, (trace metal basis)
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N Synonym: gallium, elemental,metal,unii-ch46oc8yv4,ch46oc8yv4,ingot,pellets, 6mm dia,galio,metal, packaged in poylethylene bottle,gallium, ion ga1+,atom PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis)
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N Synonym: gallium, elemental,metal,unii-ch46oc8yv4,ch46oc8yv4,ingot,pellets, 6mm dia,galio,metal, packaged in poylethylene bottle,gallium, ion ga1+,atom PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium pellets, 6mm dia, 99.99999% (metals basis)
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N Synonym: gallium, elemental,metal,unii-ch46oc8yv4,ch46oc8yv4,ingot,pellets, 6mm dia,galio,metal, packaged in poylethylene bottle,gallium, ion ga1+,atom PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium, 99.9999%, (trace metal basis)
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N Synonym: gallium, elemental,metal,unii-ch46oc8yv4,ch46oc8yv4,ingot,pellets, 6mm dia,galio,metal, packaged in poylethylene bottle,gallium, ion ga1+,atom PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium ingot, 99.999% (metals basis)
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N Synonym: gallium, elemental,metal,unii-ch46oc8yv4,ch46oc8yv4,ingot,pellets, 6mm dia,galio,metal, packaged in poylethylene bottle,gallium, ion ga1+,atom PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium metal, packaged in poylethylene bottle, 99.999% (metals basis)
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N Synonym: gallium, elemental,metal,unii-ch46oc8yv4,ch46oc8yv4,ingot,pellets, 6mm dia,galio,metal, packaged in poylethylene bottle,gallium, ion ga1+,atom PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N Synonym: gallium, elemental,metal,unii-ch46oc8yv4,ch46oc8yv4,ingot,pellets, 6mm dia,galio,metal, packaged in poylethylene bottle,gallium, ion ga1+,atom PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium, plasma standard solution, Specpure™ Ga 1000μg/mL
CAS: 7440-55-3 Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium pellets, 99.9999% (metals basis)
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N Synonym: gallium, elemental,metal,unii-ch46oc8yv4,ch46oc8yv4,ingot,pellets, 6mm dia,galio,metal, packaged in poylethylene bottle,gallium, ion ga1+,atom PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium ingot, 99.99% (metals basis)
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N Synonym: gallium, elemental,metal,unii-ch46oc8yv4,ch46oc8yv4,ingot,pellets, 6mm dia,galio,metal, packaged in poylethylene bottle,gallium, ion ga1+,atom PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium ingot, 99.99999% (metals basis)
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N Synonym: gallium, elemental,metal,unii-ch46oc8yv4,ch46oc8yv4,ingot,pellets, 6mm dia,galio,metal, packaged in poylethylene bottle,gallium, ion ga1+,atom PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-55-3 Molecular Formula: Ga Molecular Weight (g/mol): 69.72 MDL Number: MFCD00134045 InChI Key: GYHNNYVSQQEPJS-UHFFFAOYSA-N PubChem CID: 5360835 ChEBI: CHEBI:49631 IUPAC Name: gallium SMILES: [Ga]
Gallium Standard: Ga @ 1000 μg/mL in 5% HNO3, 0.5% HCl, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2