CAS RN 7440-54-2
Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium powder, -200 mesh, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium foil. 0.075mm (0.003in) thick, REacton™, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium foil, 0.62mm (0.024in) thick, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium foil, 0.25mm (0.01in) thick, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium powder, -40 mesh, 99% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium pieces, 20mm (0.79in) & down, 99.9% (REO)
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N Synonym: atom,gadolinio,unii-au0v1lm3jt,trihydride,au0v1lm3jt,foil, 3n,gadolinium, elemental,ingot,gadolinium, chips,compounds PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-54-2 Molecular Formula: Gd Molecular Weight (g/mol): 157.25 MDL Number: MFCD00011022 InChI Key: UIWYJDYFSGRHKR-UHFFFAOYSA-N PubChem CID: 23982 ChEBI: CHEBI:33375 IUPAC Name: gadolinium SMILES: [Gd]
Gadolinium Standard: Gd @ 10000 μg/mL in 5% HNO3
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2