accessibility menu, dialog, popup

UserName

Cr

CAS RN 7440-47-3

IUPAC Name: chromium
Synonyms: chromium chrom cromo chromic acid and its salts zinc chromates including zinc potassium chromate chrome chromium atom
Molecular Weight (g/mol): 52
Molecular Formula: Cr
InChi Key: VYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES: [Cr]
Molecular Weight (g/mol) 
  • (23)
  • (62)
Quantity 
  • (4)
  • (3)
  • (5)
  • (10)
  • (2)
  • (1)
  • (3)
  • (6)
  • (1)
  • (1)
  • (9)
  • (8)
Form 
  • (7)
  • (2)
  • (3)
  • (1)
  • (2)
  • (7)
  • (20)
Solvent or Matrix 
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (11)
  • (9)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (7)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (7)
  • (4)
  • (51)
  • (25)
  • (23)
  • (62)
  • (4)
  • (3)
  • (5)
  • (10)
  • (2)
  • (1)
  • (3)
  • (6)
  • (1)
  • (1)
  • (9)
  • (8)
  • (7)
  • (2)
  • (3)
  • (1)
  • (2)
  • (7)
  • (20)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (11)
  • (9)
  • (1)
  • (3)
  • (2)
  • (7)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (8)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (6)
  • (4)