accessibility menu, dialog, popup

UserName

B

CAS RN 7440-42-8

IUPAC Name: boron
Synonyms: boron boron atom boracium Bor boro bore
Molecular Weight (g/mol): 10.81
Molecular Formula: B
InChi Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N
SMILES: [B]
Solvent or Matrix 
  • (1)
  • (1)
  • (4)
  • (1)
  • (7)
  • (1)
  • (2)
  • (2)
  • (15)
Concentration 
  • (1)
  • (1)
  • (8)
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (9)
  • (2)
  • (14)
  • (25)
  • (1)
  • (1)
  • (4)
  • (1)
  • (7)
  • (1)
  • (2)
  • (2)
  • (15)
  • (1)
  • (1)
  • (8)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (6)
  • (1)
  • (3)
  • (2)