accessibility menu, dialog, popup

UserName

Tm

CAS RN 7440-30-4

IUPAC Name: thulium
Synonyms: thulium thulium atom tulio
Molecular Weight (g/mol): 168.93
Molecular Formula: Tm
InChi Key: FRNOGLGSGLTDKL-UHFFFAOYSA-N
SMILES: [Tm]
Molecular Weight (g/mol) 
  • (15)
  • (7)
Solvent or Matrix 
  • (4)
  • (7)
  • (3)
Concentration 
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
Form 
  • (4)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (7)
  • (4)
  • (15)
  • (7)
  • (4)
  • (7)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)