accessibility menu, dialog, popup

UserName

Rb

CAS RN 7440-17-7

IUPAC Name: rubidium
Synonyms: rubidium rubidium atom rubidio
Molecular Weight (g/mol): 85.47
Molecular Formula: Rb
InChi Key: IGLNJRXAVVLDKE-UHFFFAOYSA-N
SMILES: [Rb]
Molecular Weight (g/mol) 
  • (5)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Solvent or Matrix 
  • (1)
  • (5)
Concentration 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results