accessibility menu, dialog, popup

UserName

Re

CAS RN 7440-15-5

IUPAC Name: rhenium
Synonyms: rhenium rhenium atom
Molecular Weight (g/mol): 186.21
Molecular Formula: Re
InChi Key: WUAPFZMCVAUBPE-UHFFFAOYSA-N
SMILES: [Re]
Molecular Weight (g/mol) 
  • (7)
  • (35)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
Form 
  • (9)
  • (6)
  • (2)
  • (11)
Percent Purity 
  • (13)
  • (2)
  • (2)
Solvent or Matrix 
  • (1)
  • (7)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (32)
  • (8)
  • (7)
  • (35)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (9)
  • (6)
  • (2)
  • (11)
  • (13)
  • (2)
  • (2)
  • (1)
  • (7)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)