CAS RN 7439-93-2
Lithium, 99+%, granular, dry
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N Synonym: litio,litium,lithium, metallic,lithium, elemental,monohydride,unii-9fn79x2m3f,hsdb 647,hydride lih,hydrure de french,hsdb 549 PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium wire, 3.2mm (0.125in) dia, 99.8%, Na 0.03% max
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N Synonym: litio,litium,lithium, metallic,lithium, elemental,monohydride,unii-9fn79x2m3f,hsdb 647,hydride lih,hydrure de french,hsdb 549 PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl
CAS: 7439-93-2 | Li | 6.94 g/mol
Lithium granules, 1-6mm (0.04-0.2in), 99%
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N Synonym: litio,litium,lithium, metallic,lithium, elemental,monohydride,unii-9fn79x2m3f,hsdb 647,hydride lih,hydrure de french,hsdb 549 PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
CAS: 7439-93-2 Molecular Formula: HNO3 Molecular Weight (g/mol): 63.01 MDL Number: MFCD00011349 InChI Key: GRYLNZFGIOXLOG-UHFFFAOYSA-N PubChem CID: 944 ChEBI: CHEBI:48107 IUPAC Name: nitric acid SMILES: O[N+]([O-])=O
Lithium Standard For ICP, 1000μg/mL in 2% HNO3 Assurance Grade, SPEX CertiPrep™
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium Standard: Li+ @ 1000 μg/mL in H2O, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Multi-Cation Standard 1: Li+ @ 50; Na+ @ 200; Mg+2, NH4+ @ 250; Ca+2, K+ @ 500 μg/mL in dil. HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lithium AA Standard: Li @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lithium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]