CAS RN 7439-89-6
Iron granules, 1-2mm (0.04-0.08in), 99.98% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron, 99%, powder, -70 mesh (<212 micron)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3
CAS: 7439-89-6 | Fe | 55.85 g/mol
Iron powder, spherical, APS 6-10 micron, reduced, 99.5%, Thermo Scientific Chemicals
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron powder, spherical, <20 micron, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron foil, 0.5mm (0.02in) thick, 99.99% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron rod, 5mm (0.2in) dia, 99.99% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron pieces, irregularly shaped, 25mm (1.0in) & down, 99.97+% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron foil, 2.0mm (0.08in) thick, Puratronic™, 99.995% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron foil, 1.0mm (0.04in) thick, Puratronic™, 99.995% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron sponge, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Iron wire, 0.075mm (0.003in) dia, Puratronic™, 99.995% (metals basis)
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.85 MDL Number: MFCD00010999 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N Synonym: ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]