accessibility menu, dialog, popup

UserName

CAS RN 7429-37-0

IUPAC Name: (1R,2R)-1,2-dibromocyclohexane
Synonyms: (1R,2R)-1,2-dibromocyclohexane
Molecular Weight (g/mol): 241.95
Molecular Formula: C6H10Br2
InChi Key: CZNHKZKWKJNOTE-PHDIDXHHSA-N
SMILES: Br[C@@H]1CCCC[C@H]1Br
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)