accessibility menu, dialog, popup

UserName

H3COFO

CAS RN 74266-68-5

IUPAC Name: 3-fluoro-2-methoxybenzaldehyde
Synonyms: 3-fluoro-2-methoxybenzaldehyde
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: UMLOYMZBAUIGKF-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results