accessibility menu, dialog, popup

UserName

CAS RN 7400-27-3

IUPAC Name: hydrogen tert-butylhydrazine chloride
Synonyms: hydrogen tert-butylhydrazine chloride
Molecular Weight (g/mol): 124.61
Molecular Formula: C4H13ClN2
InChi Key: DDPWVABNMBRBFI-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)(C)NN
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results