accessibility menu, dialog, popup

UserName

CAS RN 74-88-4

IUPAC Name: iodomethane
Synonyms: methyl iodide 14C-labeled methyl iodide 14c-labeled methyl iodide 132I-labeled methyl iodide methyl iodide, 132i-labeled 132i-labeled methyl iodide methyl iodide, 13C-labeled jod-methan 11C-labeled methyl iodide MeI
Molecular Weight (g/mol): 141.94
Molecular Formula: CH3I
InChi Key: INQOMBQAUSQDDS-UHFFFAOYSA-N
SMILES: CI
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (7)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (7)
  • (2)
  • (2)
  • (1)