accessibility menu, dialog, popup

UserName

CH3BH3COH3C

CAS RN 7397-46-8

IUPAC Name: methyl diethylborinate
Synonyms: methyl diethylborinate
Molecular Weight (g/mol): 99.97
Molecular Formula: C5H13BO
InChi Key: FESAXEDIWWXCNG-UHFFFAOYSA-N
SMILES: CCB(CC)OC
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)